siRNA Details
| Gene Symbol: Ubox5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033997.1 | NP_001029169.1 | 4 | 1298 | ||
| XM_008762160.1 | XP_008760382.1 | 3 | 1209 | |||
| XM_008762161.1 | XP_008760383.1 | 3 | 1209 | |||
| GenBank mRNA | BC101912.1 | - | 4 | 1298 | 7933511 | |

Ubox5
Genomic Map