siRNA Details
| Gene Symbol: Pank2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106513.1 | NP_001099983.1 | 7 | 1347 | ||
| XM_008762162.1 | XP_008760384.1 | 7 | 1292 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pank2| Gene Symbol: Pank2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106513.1 | NP_001099983.1 | 7 | 1347 | ||
| XM_008762162.1 | XP_008760384.1 | 7 | 1292 | |||