siRNA Details
| Gene Symbol: Fermt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106515.1 | NP_001099985.1 | 10 | 1751 | ||
| XM_006235076.2 | XP_006235138.1 | 10 | 1525 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fermt1| Gene Symbol: Fermt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106515.1 | NP_001099985.1 | 10 | 1751 | ||
| XM_006235076.2 | XP_006235138.1 | 10 | 1525 | |||