siRNA Details
| Gene Symbol: Tmx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100529.1 | NP_001093999.1 | 8 | 1066 | ||
| GenBank mRNA | BC094304.1 | - | 5 | 498 | ||
| BC109379.1 | - | 5 | 498 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmx4| Gene Symbol: Tmx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100529.1 | NP_001093999.1 | 8 | 1066 | ||
| GenBank mRNA | BC094304.1 | - | 5 | 498 | ||
| BC109379.1 | - | 5 | 498 | |||