siRNA Details
| Gene Symbol: Ankef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106516.1 | NP_001099986.1 | 8 | 2240 | ||
| XM_008762252.1 | XP_008760474.1 | 8 | 3952 | |||
| GenBank mRNA | BC160831.1 | - | 8 | 1390 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ankef1| Gene Symbol: Ankef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106516.1 | NP_001099986.1 | 8 | 2240 | ||
| XM_008762252.1 | XP_008760474.1 | 8 | 3952 | |||
| GenBank mRNA | BC160831.1 | - | 8 | 1390 | ||