siRNA Details
| Gene Symbol: Snx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106518.1 | NP_001099988.1 | 10 | 953 | ||
| XM_006235119.2 | XP_006235181.1 | 10 | 1003 | |||
| GenBank mRNA | BC160883.1 | - | 10 | 934 | 7382263 | |
| FQ225134.1 | - | 10 | 934 | |||
| FQ231966.1 | - | 10 | 936 | |||

Snx5
Genomic Map