siRNA Details
| Gene Symbol: Tm9sf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025649.1 | NP_001020820.1 | 18 | 2106 | ||
| XM_006235284.2 | XP_006235346.1 | 18 | 2124 | |||
| GenBank mRNA | BC099133.1 | - | 18 | 2106 | 7454474 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tm9sf4| Gene Symbol: Tm9sf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025649.1 | NP_001020820.1 | 18 | 2106 | ||
| XM_006235284.2 | XP_006235346.1 | 18 | 2124 | |||
| GenBank mRNA | BC099133.1 | - | 18 | 2106 | 7454474 | |