siRNA Details
| Gene Symbol: Sun5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106530.1 | NP_001100000.1 | 9 | 710 | ||
| XM_008762312.1 | XP_008760534.1 | 11 | 881 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sun5| Gene Symbol: Sun5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106530.1 | NP_001100000.1 | 9 | 710 | ||
| XM_008762312.1 | XP_008760534.1 | 11 | 881 | |||