siRNA Details
| Gene Symbol: Gria3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001112742.1 | NP_001106213.1 | 2 | 196 | ||
| NM_032990.2 | NP_116785.2 | 2 | 196 | |||
| XM_006257496.2 | XP_006257558.1 | 3 | 273 | |||
| XM_006257498.2 | XP_006257560.1 | 3 | 296 | |||
| XM_006257499.2 | XP_006257561.1 | 2 | 325 | |||
| XM_006257501.2 | XP_006257563.1 | 3 | 268 | |||
| XM_008773512.1 | XP_008771734.1 | 2 | 325 | |||
| GenBank mRNA | M36420.1 | - | 2 | 140 | ||
| M38062.1 | - | 2 | 140 | |||
| M85036.2 | - | 3 | 306 | |||
| X54656.1 | - | 2 | 196 | |||

Gria3
Genomic Map