siRNA Details
Gene Symbol: Cnbd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762318.1 | XP_008760540.1 | 14 | 2125 | ||
XM_008762319.1 | XP_008760541.1 | 13 | 2068 | |||
XM_008762320.1 | XP_008760542.1 | 12 | 2098 | |||
XM_008762321.1 | XP_008760543.1 | 14 | 2145 |
