siRNA Details
Gene Symbol: Cnbd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762318.1 | XP_008760540.1 | 10, 11 | 1367 | ||
XM_008762319.1 | XP_008760541.1 | 9, 10 | 1310 | |||
XM_008762320.1 | XP_008760542.1 | 8, 9 | 1340 | |||
XM_008762321.1 | XP_008760543.1 | 10, 11 | 1367 | |||
GenBank mRNA | AY539878.1 | - | 10, 11 | 1595 |
