siRNA Details
| Gene Symbol: Serinc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008312.1 | NP_001008313.1 | 9, 10 | 1383 | ||
| GenBank mRNA | BC085853.1 | - | 9, 10 | 1383 | 7190015 | |
| EU220431.1 | - | 9, 10 | 1329 | |||
| FQ213438.1 | - | 1, 2 | 244 | |||
| FQ222836.1 | - | 3, 4 | 466 | |||

Serinc3
Genomic Map