siRNA Details
| Gene Symbol: Ctsa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011959.1 | NP_001011959.1 | 9 | 1036 | ||
| XM_006235534.1 | XP_006235596.1 | 10 | 1294 | |||
| GenBank mRNA | BC078934.1 | - | 9 | 1036 | 7108104 | |
| FQ219992.1 | - | 2 | 89 | |||
| FQ229081.1 | - | 9 | 1190 | |||

Ctsa
Genomic Map