siRNA Details
| Gene Symbol: Ctsa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011959.1 | NP_001011959.1 | 1 | 247 | ||
| XM_006235534.1 | XP_006235596.1 | 2 | 505 | |||
| GenBank mRNA | BC078934.1 | - | 1 | 247 | 7108104 | |
| FQ229081.1 | - | 2 | 288 | |||

Ctsa
Genomic Map