siRNA Details
| Gene Symbol: Hmgcs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017268.1 | NP_058964.1 | 3 | 440 | ||
| XM_006231959.1 | XP_006232021.1 | 2 | 609 | |||
| GenBank mRNA | AY724522.1 | - | 2 | 426 | ||
| FQ211444.1 | - | 2 | 412 | |||
| X52625.1 | - | 3 | 440 | |||

Hmgcs1
Genomic Map