siRNA Details
| Gene Symbol: Arfgef2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181083.2 | NP_851597.2 | 37 | 5009 | ||
| XM_006235640.2 | XP_006235702.1 | 37 | 5191 | |||
| GenBank mRNA | AY255526.2 | - | 37 | 5009 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arfgef2| Gene Symbol: Arfgef2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181083.2 | NP_851597.2 | 37 | 5009 | ||
| XM_006235640.2 | XP_006235702.1 | 37 | 5191 | |||
| GenBank mRNA | AY255526.2 | - | 37 | 5009 | ||