siRNA Details
| Gene Symbol: Slc38a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181090.2 | NP_851604.1 | 4 | 661 | ||
| GenBank mRNA | AF249673.1 | - | 4 | 661 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc38a2| Gene Symbol: Slc38a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181090.2 | NP_851604.1 | 4 | 661 | ||
| GenBank mRNA | AF249673.1 | - | 4 | 661 | ||