siRNA Details
| Gene Symbol: Slc38a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181090.2 | NP_851604.1 | 9 | 1071 | ||
| XM_006242282.1 | XP_006242344.1 | 9 | 1111 | |||
| XM_006242283.1 | XP_006242345.1 | 8 | 995 | |||
| GenBank mRNA | AF173682.2 | - | 9 | 872 | ||
| AF249673.1 | - | 9 | 1071 | |||
| AF273024.1 | - | 9 | 906 | |||

Slc38a2
Genomic Map