siRNA Details
| Gene Symbol: RGD1565819 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171096.1 | NP_001164567.1 | 1 | 3442 | ||
| XM_006235714.2 | XP_006235776.1 | 2 | 3872 | |||
| XM_008762455.1 | XP_008760677.1 | 2 | 4600 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1565819| Gene Symbol: RGD1565819 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171096.1 | NP_001164567.1 | 1 | 3442 | ||
| XM_006235714.2 | XP_006235776.1 | 2 | 3872 | |||
| XM_008762455.1 | XP_008760677.1 | 2 | 4600 | |||