siRNA Details
| Gene Symbol: Hat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009657.1 | NP_001009657.1 | 5 | 380 | ||
| GenBank mRNA | BC087663.1 | - | 5 | 380 | 7315259 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hat1| Gene Symbol: Hat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009657.1 | NP_001009657.1 | 5 | 380 | ||
| GenBank mRNA | BC087663.1 | - | 5 | 380 | 7315259 | |