siRNA Details
| Gene Symbol: C8g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106555.1 | NP_001100025.1 | 5 | 529 | ||
| XM_006233581.2 | XP_006233643.1 | 5 | 914 | |||
| GenBank mRNA | FQ210613.1 | - | 5 | 489 | ||
| FQ212796.1 | - | 4 | 668 | |||
| FQ218819.1 | - | 5 | 489 | |||

C8g
Genomic Map