siRNA Details
| Gene Symbol: Anapc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100532.1 | NP_001094002.1 | 3 | 889 | ||
| GenBank mRNA | BC166796.1 | - | 3 | 889 | 7938000 | |
| FQ224461.1 | - | 3 | 899 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Anapc2| Gene Symbol: Anapc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100532.1 | NP_001094002.1 | 3 | 889 | ||
| GenBank mRNA | BC166796.1 | - | 3 | 889 | 7938000 | |
| FQ224461.1 | - | 3 | 899 | |||