siRNA Details
| Gene Symbol: Npdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004231.1 | NP_001004231.1 | 6 | 772 | ||
| XM_006233584.2 | XP_006233646.1 | 5 | 1410 | |||
| GenBank mRNA | BC079156.1 | - | 6 | 772 | 7128989 | |
| FQ214366.1 | - | 6 | 616 | |||

Npdc1
Genomic Map