siRNA Details
| Gene Symbol: Slc2a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106562.1 | NP_001100032.1 | 8 | 1255 | ||
| XM_006233757.2 | XP_006233819.1 | 8 | 1246 | |||
| XM_008761634.1 | XP_008759856.1 | 8 | 1162 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc2a6| Gene Symbol: Slc2a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106562.1 | NP_001100032.1 | 8 | 1255 | ||
| XM_006233757.2 | XP_006233819.1 | 8 | 1246 | |||
| XM_008761634.1 | XP_008759856.1 | 8 | 1162 | |||