siRNA Details
| Gene Symbol: Tmem8c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134517.1 | NP_001127989.1 | 2 | 374 | ||
| XM_008761635.1 | XP_008759857.1 | 2 | 376 | |||
| XM_008761636.1 | XP_008759858.1 | 2 | 125 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem8c| Gene Symbol: Tmem8c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134517.1 | NP_001127989.1 | 2 | 374 | ||
| XM_008761635.1 | XP_008759857.1 | 2 | 376 | |||
| XM_008761636.1 | XP_008759858.1 | 2 | 125 | |||