siRNA Details
Gene Symbol: Tmem8c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134517.1 | NP_001127989.1 | 1 | 295 | ||
XM_008761635.1 | XP_008759857.1 | 1 | 297 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmem8c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134517.1 | NP_001127989.1 | 1 | 295 | ||
XM_008761635.1 | XP_008759857.1 | 1 | 297 |