siRNA Details
| Gene Symbol: Med27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106565.1 | NP_001100035.1 | 3, 4 | 515 | ||
| XM_008761638.1 | XP_008759860.1 | 3, 4 | 521 | |||
| XM_008761639.1 | XP_008759861.1 | 2, 3 | 554 | |||
| GenBank mRNA | BC167032.1 | - | 3, 4 | 490 | 9092805 | |

Med27
Genomic Map