siRNA Details
| Gene Symbol: Med27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106565.1 | NP_001100035.1 | 8 | 947 | ||
| XM_008761639.1 | XP_008759861.1 | 7 | 986 | |||
| GenBank mRNA | BC167032.1 | - | 8 | 922 | 9092805 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Med27| Gene Symbol: Med27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106565.1 | NP_001100035.1 | 8 | 947 | ||
| XM_008761639.1 | XP_008759861.1 | 7 | 986 | |||
| GenBank mRNA | BC167032.1 | - | 8 | 922 | 9092805 | |