siRNA Details
| Gene Symbol: Fam73b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106566.1 | NP_001100036.1 | 2 | 256 | ||
| XM_006233781.1 | XP_006233843.1 | 2 | 305 | |||
| XM_006233782.2 | XP_006233844.1 | 2 | 321 | |||
| GenBank mRNA | FQ212119.1 | - | 2 | 224 | ||
| FQ231217.1 | - | 2 | 260 | |||

Fam73b
Genomic Map