siRNA Details
| Gene Symbol: Mapkap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011964.1 | NP_001011964.1 | 5, 6 | 1105 | ||
| XM_006233949.2 | XP_006234011.1 | 6, 7 | 1291 | |||
| XM_006233950.2 | XP_006234012.1 | 5, 6 | 843 | |||
| XM_008761720.1 | XP_008759942.1 | 5, 6 | 890 | |||
| XM_008761721.1 | XP_008759943.1 | 4, 5 | 805 | |||
| XM_008761722.1 | XP_008759944.1 | 3, 4 | 715 | |||
| XM_008761723.1 | XP_008759945.1 | 3, 4 | 393 | |||
| GenBank mRNA | BC079073.1 | - | 5, 6 | 1105 | 7122012 | |

Mapkap1
Genomic Map