siRNA Details
| Gene Symbol: Psmd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106569.2 | NP_001100039.2 | 10 | 1307 | ||
| XM_006233969.2 | XP_006234031.1 | 10 | 1836 | |||
| XM_006233970.2 | XP_006234032.1 | 11 | 1926 | |||
| GenBank mRNA | BC166850.1 | - | 10 | 1289 | 9088649 | |

Psmd5
Genomic Map