siRNA Details
| Gene Symbol: Phf19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106570.1 | NP_001100040.1 | 11, 12 | 1167 | ||
| XM_006233971.2 | XP_006234033.1 | 11, 12 | 1229 | |||
| XM_008761740.1 | XP_008759962.1 | 6, 7 | 970 | |||

Phf19
Genomic Map