siRNA Details
| Gene Symbol: Phf19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106570.1 | NP_001100040.1 | 15 | 1756 | ||
| XM_006233971.2 | XP_006234033.1 | 15 | 1818 | |||
| XM_008761740.1 | XP_008759962.1 | 10 | 1559 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Phf19| Gene Symbol: Phf19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106570.1 | NP_001100040.1 | 15 | 1756 | ||
| XM_006233971.2 | XP_006234033.1 | 15 | 1818 | |||
| XM_008761740.1 | XP_008759962.1 | 10 | 1559 | |||