siRNA Details
| Gene Symbol: Olr424 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000392.1 | NP_001000392.1 | 1 | 657 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr424| Gene Symbol: Olr424 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000392.1 | NP_001000392.1 | 1 | 657 | ||