siRNA Details
| Gene Symbol: Olfml2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106572.1 | NP_001100042.1 | 6 | 1211 | ||
| XM_006234074.2 | XP_006234136.1 | 7 | 1419 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfml2a| Gene Symbol: Olfml2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106572.1 | NP_001100042.1 | 6 | 1211 | ||
| XM_006234074.2 | XP_006234136.1 | 7 | 1419 | |||