siRNA Details
| Gene Symbol: Pus7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170589.1 | NP_001164060.1 | 16 | 2044 | ||
| XM_008762634.1 | XP_008760856.1 | 16 | 2176 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pus7| Gene Symbol: Pus7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170589.1 | NP_001164060.1 | 16 | 2044 | ||
| XM_008762634.1 | XP_008760856.1 | 16 | 2176 | |||