siRNA Details
| Gene Symbol: Napepld | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199381.1 | NP_955413.1 | 2 | 713 | ||
| XM_006235908.2 | XP_006235970.1 | 3 | 2135 | |||
| XM_006235910.2 | XP_006235972.1 | 3 | 982 | |||
| XM_006235911.2 | XP_006235973.1 | 3 | 936 | |||
| XM_008762638.1 | XP_008760860.1 | 3 | 4384 | |||
| XM_008762639.1 | XP_008760861.1 | 3 | 1645 | |||
| GenBank mRNA | AB112351.1 | - | 2 | 713 | ||

Napepld
Genomic Map