siRNA Details
| Gene Symbol: Psmc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031595.1 | NP_113783.1 | 6 | 741 | ||
| GenBank mRNA | BC062019.1 | - | 6 | 651 | 5620971 | |
| D83522.1 | - | 6 | 615 | |||
| FQ225567.1 | - | 5 | 400 | |||
| FQ225947.1 | - | 5 | 400 | |||
| FQ226273.1 | - | 5 | 400 | |||
| FQ232987.1 | - | 5 | 400 | |||
| FQ233836.1 | - | 5 | 400 | |||
| U77918.1 | - | 6 | 741 | |||

Psmc3
Genomic Map