siRNA Details
| Gene Symbol: LOC102551166 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006235984.2 | XP_006236046.1 | 6, 7 | 957 | ||
| XM_006235985.2 | XP_006236047.1 | 6, 7 | 957 | |||
| XM_006235986.2 | XP_006236048.1 | 6, 7 | 957 | |||
| XM_006235987.2 | XP_006236049.1 | 2, 3 | 187 | |||

LOC102551166
Genomic Map