siRNA Details
| Gene Symbol: Sema3e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106579.1 | NP_001100049.1 | 10, 11 | 1743 | ||
| XM_008762698.1 | XP_008760920.1 | 9, 10 | 1511 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sema3e| Gene Symbol: Sema3e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106579.1 | NP_001100049.1 | 10, 11 | 1743 | ||
| XM_008762698.1 | XP_008760920.1 | 9, 10 | 1511 | |||