siRNA Details
| Gene Symbol: Cyp11a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017286.2 | NP_058982.1 | 7 | 1204 | ||
| GenBank mRNA | BC089100.1 | - | 7 | 1229 | 7307104 | |
| J05156.1 | - | 7 | 1224 | |||
| M22615.1 | - | 2 | 175 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp11a1| Gene Symbol: Cyp11a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017286.2 | NP_058982.1 | 7 | 1204 | ||
| GenBank mRNA | BC089100.1 | - | 7 | 1229 | 7307104 | |
| J05156.1 | - | 7 | 1224 | |||
| M22615.1 | - | 2 | 175 | |||