siRNA Details
| Gene Symbol: Kcnq3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031597.4 | NP_113785.3 | 7 | 1488 | ||
| XM_008765495.1 | XP_008763717.1 | 7 | 816 | |||
| GenBank mRNA | AF091247.1 | - | 8 | 1034 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnq3| Gene Symbol: Kcnq3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031597.4 | NP_113785.3 | 7 | 1488 | ||
| XM_008765495.1 | XP_008763717.1 | 7 | 816 | |||
| GenBank mRNA | AF091247.1 | - | 8 | 1034 | ||