siRNA Details
| Gene Symbol: Lrrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_216085.7 | XP_216085.3 | 2 | 2208 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrd1| Gene Symbol: Lrrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_216085.7 | XP_216085.3 | 2 | 2208 | ||