siRNA Details
| Gene Symbol: Mcm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017287.1 | NP_058983.1 | 7 | 1070 | ||
| GenBank mRNA | BC166822.1 | - | 7 | 990 | 8369193 | |
| CD371511.1 | - | 3 | 348 | |||
| U17565.1 | - | 1 | 20 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mcm6| Gene Symbol: Mcm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017287.1 | NP_058983.1 | 7 | 1070 | ||
| GenBank mRNA | BC166822.1 | - | 7 | 990 | 8369193 | |
| CD371511.1 | - | 3 | 348 | |||
| U17565.1 | - | 1 | 20 | |||