siRNA Details
| Gene Symbol: ST7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004102.1 | NP_001004102.1 | 13, 14 | 1613 | ||
| XM_006236132.2 | XP_006236194.1 | 14, 15 | 2522 | |||
| XM_006236133.2 | XP_006236195.1 | 14, 15 | 2815 | |||
| XM_006236134.2 | XP_006236196.1 | 14, 15 | 2051 | |||
| XM_006236135.2 | XP_006236197.1 | 13, 14 | 1982 | |||
| XM_008762700.1 | XP_008760922.1 | 14, 15 | 3247 | |||
| XM_008762701.1 | XP_008760923.1 | 15, 16 | 2209 | |||
| GenBank mRNA | BC078945.1 | - | 15, 16 | 1869 | ||

ST7
Genomic Map