siRNA Details
| Gene Symbol: ST7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004102.1 | NP_001004102.1 | 7, 8 | 983 | ||
| XM_006236132.2 | XP_006236194.1 | 8, 9 | 1892 | |||
| XM_006236133.2 | XP_006236195.1 | 8, 9 | 2185 | |||
| XM_006236134.2 | XP_006236196.1 | 8, 9 | 1421 | |||
| XM_006236135.2 | XP_006236197.1 | 7, 8 | 1352 | |||
| XM_008762700.1 | XP_008760922.1 | 8, 9 | 2617 | |||
| XM_008762701.1 | XP_008760923.1 | 9, 10 | 1579 | |||
| GenBank mRNA | BC078945.1 | - | 9, 10 | 1239 | ||
| BC099820.1 | - | 7, 8 | 1065 | 7930680 | ||

ST7
Genomic Map