siRNA Details
| Gene Symbol: Gabrr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017291.3 | NP_058987.2 | 5, 6 | 690 | ||
| XM_008763609.1 | XP_008761831.1 | 5, 6 | 473 | |||
| GenBank mRNA | U21070.1 | - | 5, 6 | 647 | ||
| X95579.1 | - | 5, 6 | 690 | |||

Gabrr1
Genomic Map