siRNA Details
| Gene Symbol: Tnpo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106587.1 | NP_001100057.1 | 22 | 3106 | ||
| XM_006236186.2 | XP_006236248.1 | 23 | 2880 | |||
| GenBank mRNA | AY724521.1 | - | 22 | 2880 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_177296, BC075678, BC052756, AK011912, BC064646, and others.

Tnpo3
Genomic Map