siRNA Details
Gene Symbol: Tnpo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106587.1 | NP_001100057.1 | 17 | 2501 | ||
XM_006236186.2 | XP_006236248.1 | 18 | 2275 | |||
GenBank mRNA | AY724521.1 | - | 17 | 2275 |
