siRNA Details
| Gene Symbol: Gabrr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017292.2 | NP_058988.1 | 6 | 780 | ||
| XM_008763610.1 | XP_008761832.1 | 8 | 766 | |||
| XM_008763611.1 | XP_008761833.1 | 5 | 1712 | |||
| GenBank mRNA | D38494.1 | - | 6 | 780 | ||

Gabrr2
Genomic Map